Få mere at vide
Ethyl 4-(1-Hydroxy-1-methylethyl)-2-propyl-1H-imidazole-5-carboxylate, Mikromol™
Artikelnummer. 30050158
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050158 100 mg
30050917 25 mg
30458242 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050158 Leverandør Mikromol Leverandørnr. MM1317.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 4-(1-Hydroxy-1-methylethyl)-2-propyl-1H-imidazole-5-carboxylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 4-(1-Hydroxy-1-methylethyl)-2-propyl-1H-imidazole-5-carboxylate
CAS 144689-93-0
Analyt- eller komponentnavne Ethyl 4-(1-Hydroxy-1-methylethyl)-2-propyl-1H-imidazole-5-carboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C12 H20 N2 O3
Certificeringer/compliance ISO 9001
Synonym 2-[4-[2-(3-Methyl-1,1-dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine Maleate, 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylethyl)-2-propyl-, ethyl ester, 1H-Imidazole-4-carboxylic acid, 5-(1-hydroxy-1-methylethyl)-2-propyl-, ethyl ester (9CI), 2-Propyl-5-(1-hydroxy-1-methylethyl)-3H-imidazole-4-carboxylic acid ethyl ester, 4-(1-Hydroxy-1-methylethyl)-2-propylimidazole-5-carboxylic acid ethyl ester, Ethyl 4-(1-hydroxy-1-methylethyl)-2-propylimidazole-5-carboxylate
SMIL CCCc1nc(c([nH]1)C(=O)OCC)C(C)(C)O
Molekylvægt (g/mol) 240.3
Mængde 100 mg
Formel vægt 240.303 g/mol