Få mere at vide
Ethyl 3-(p-Menthane-3-carboxamido)acetate, TRC
Artikelnummer. 30136217
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136217 5 g
30133603 10 g
30011940 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136217 Leverandør Toronto Research Chemicals Leverandørnr. TRCE6848905G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68489-14-5
Molekylær formel C15 H27 N O3
Molekylvægt (g/mol) 269.38
Synonym N-[[(1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl]carbonyl]glycine Ethyl Ester, CPS 337, FEMA 4309;
IUPAC navn ethyl 2-[[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexanecarbonyl]amino]acetate
SMIL CCOC(=O)CNC(=O)[C@@H]1C[C@H](C)CC[C@H]1C(C)C
Tekniske data
Kemisk navn eller materiale Ethyl 3-(p-Menthane-3-carboxamido)acetate
Renhedsgrad noter HPLC
InChI formel InChI=1S/C15H27NO3/c1-5-19-14(17)9-16-15(18)13-8-11(4)6-7-12(13)10(2)3/h10-13H,5-9H2,1-4H3,(H,16,18)/t11-,12+,13-/m1/s1
Mængde 5 g
Formel vægt 269.199
Procent renhed >95%