Få mere at vide
Ethyl 3-Oxo-4-aza-5Alpha-androst-1-ene-17Beta-carboxylate, TRC
Artikelnummer. 30134165
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134165 1 g
17912245 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134165 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9253451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 157307-36-3
Molekylær formel C21 H31 N O3
Molekylvægt (g/mol) 345.48
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, ethyl ester, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, : 4-Azaandrost-1-ene-17-carboxylic acid, 3-oxo-, ethyl ester, (5α,17β)-, 1H-Indeno[5,4-f]quinoline, 4-azaandrost-1-ene-17-carboxylic acid deriv., Ethyl 3-oxo-4-aza-5α-androst-1-ene-17β-carboxylate, 3-Oxo-4-aza-5α-androst-1-ene-17β-carboxylic Acid Ethyl Ester, Dutasteride Imp C (EP), Dutasteride Ethyl Ester
IUPAC navn ethyl (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylate
SMIL CCOC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale Ethyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate
Mængde 1 g
InChI formel InChI=1S/C21H31NO3/c1-4-25-19(24)16-7-6-14-13-5-8-17-21(3,12-10-18(23)22-17)15(13)9-11-20(14,16)2/h10,12-17H,4-9,11H2,1-3H3,(H,22,23)/t13-,14-,15-,16+,17+,20-,21+/m0/s1
Formel vægt 345.2304