Learn More
Ethyl 3-oxo-3-(o-tolyl)propanoate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 51725-82-7 |
| Molecular Formula | C12H14O3 |
| MDL Number | MFCD03424817 |
| Quantity | 5 g |
| SMILES | O=C(OCC)CC(C1=CC=CC=C1C)=O |
| Molecular Weight (g/mol) | 206.24 |
| Percent Purity | 95% |
| Chemical Name or Material | Ethyl 3-oxo-3-(o-tolyl)propanoate |