Få mere at vide
Ethyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate (3-Methyl-flavone-8-carboxylic Acid Ethyl Ester), Mikromol™
Beskrivelse
Ethyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate (3-Methyl-flavone-8-carboxylic Acid Ethyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate |
| CAS | 35888-94-9 |
| Analyt- eller komponentnavne | Ethyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H16 O4 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Flavoxate Hydrochloride Imp. B (EP), Flavoxate Imp. B (EP), Flavoxate USP Related Compound C, Flavoxate USP RC C, 3-Methyl-flavone-8-carboxylic Acid Ethyl Ester, Ethyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate, Flavoxate Related Compound C, Flavoxate Hydrochloride Impurity B, Flavoxate Impurity B |
| InChI formel | InChI=1S/C19H16O4/c1-3-22-19(21)15-11-7-10-14-16(20)12(2)17(23-18(14)15)13-8-5-4-6-9-13/h4-11H,3H2,1-2H3 |
| SMIL | CCOC(=O)c1cccc2C(=O)C(=C(Oc12)c3ccccc3)C |
| IUPAC navn | ethyl 3-methyl-4-oxo-2-phenylchromene-8-carboxylate |
| Vis mere |