Få mere at vide
Ethyl 3-Methyl-2-butenoate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 3-Methyl-2-butenoate |
| CAS | 638-10-8 |
| Molekylær formel | C7H12O2 |
| Mængde | 10 g |
| Synonym | 3-Methyl-2-butenoic Acid Ethyl Ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-Methyl-2-butenoate, Ethyl 3-Methylcrotonate, Ethyl Dimethylacrylate, Ethyl Isobutenoate, Ethyl Isopropylideneacetate, Ethyl Senecioate, Ethyl beta,beta-Dimethylacrylate, Ethyl beta-Methylcrotonate, NSC 61853, NSC 99208 |
| InChI formel | InChI=1S/C7H12O2/c1-4-9-7(8)5-6(2)3/h5H,4H2,1-3H3 |
| SMIL | CCOC(=O)C=C(C)C |
| IUPAC navn | ethyl 3-methylbut-2-enoate |
| Molekylvægt (g/mol) | 128.17 |
| Formel vægt | 128.0837 |