missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl 3-Hydroxybutyrate-d5, TRC

Artikelnummer. 30384473
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30384473 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30384473 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91895810MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl 3-Hydroxybutyrate-d5
CAS 1206486-01-2
Molekylær formel C6H7D5O3
Mængde 10 mg
Synonym 3-Hydroxy-butanoic Acid Ethyl Ester-d6, 3-hydroxy-butyric acid ethyl ester-d6, β-Hydroxy-butyric Acid Ethyl Ester-d6, (±)-3-Hydroxybutanoic acid ethyl ester-d6, (±)-Ethyl 3-hydroxybutyrate-d6, 3-Hydroxybutanoic acid ethyl ester-d6, 3-Hydroxybutyric acid ethyl ester-d6, DL-3-Hydroxy-n-butyrate ethyl ester-d6, Ethyl (±)-3-hydroxybutanoate-d6, Ethyl 3-hydroxybutanoate-d6, Ethyl DL-3-hydroxybutyrate-d6, Ethyl β-hydroxybutyrate-d6, NSC 42916-d6, NSC 8115-d6
InChI formel InChI=1S/C6H12O3/c1-3-9-6(8)4-5(2)7/h5,7H,3-4H2,1-2H3/i1D3,3D2
SMIL OC(CC(OC([2H])(C([2H])([2H])[2H])[2H])=O)C
IUPAC navn ethyl-d5 3-hydroxybutanoate
Molekylvægt (g/mol) 137.19
Formel vægt 269.19
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.