Få mere at vide
Ethyl 3-[(Diaminomethylidene)amino]-4-methylbenzoate Nitrate, Mikromol™
Beskrivelse
Ethyl 3-[(Diaminomethylidene)amino]-4-methylbenzoate Nitrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 3-[(Diaminomethylidene)amino]-4-methylbenzoate Nitrate |
| CAS | 641569-96-2 |
| Analyt- eller komponentnavne | Ethyl 3-[(Diaminomethylidene)amino]-4-methylbenzoate Nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H15 N3 O2 . H N O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzoic acid, 3-[(aminoiminomethyl)amino]-4-methyl-, ethyl ester, nitrate (1:1), Benzoic acid, 3-[(aminoiminomethyl)amino]-4-methyl-, ethyl ester, mononitrate (9CI), 3-[(Aminoiminomethyl)amino]-4-methylbenzoic acid ethyl ester mononitrate |
| InChI formel | InChI=1S/C11H15N3O2.HNO3/c1-3-16-10(15)8-5-4-7(2)9(6-8)14-11(12)13;2-1(3)4/h4-6H,3H2,1-2H3,(H4,12,13,14);(H,2,3,4) |
| SMIL | CCOC(=O)c1ccc(C)c(c1)N=C(N)N.O[N+](=O)[O-] |
| IUPAC navn | ethyl 3-(diaminomethylideneamino)-4-methylbenzoate;nitric acid |
| Vis mere |