missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl 3,5-Dimethyl-2-pyrrolecarboxylate, 98%, BLD Pharm™

Artikelnummer. 31069400
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31069400 5 g
31067068 100 g
31061381 10 g
31066405 25 g
31059970 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31069400 Leverandør BLD Pharm Leverandørnr. BD113775g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Tekniske data

Kemisk navn eller materiale Ethyl 3,5-Dimethyl-2-pyrrolecarboxylate
CAS 2199-44-2
Molekylær formel C9H13NO2
MDL nummer MFCD00010645
Mængde 5 g
SMIL O=C(C1=C(C)C=C(C)N1)OCC
Molekylvægt (g/mol) 167.21
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.