missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-bis(trifluoromethyl)phenylacetate, 98%, Thermo Scientific™

Catalog Number p-7028481
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
144632-97-3
C12H10F6O2
300.2
MFCD08689894
ICTVPOAERFKXSS-UHFFFAOYSA-N
ethyl 3,5-bis trifluoromethyl phenyl acetate, ethyl 3,5-bis trifluoromethyl phenylacetate, 3,5-bis trifluoromethyl phenylacetic acid ethyl ester, ethyl 2-3,5-bis trifluoromethyl phenyl acetate, ethyl 2-3,5-bis trifluoromethyl phenyl ethanoate, 3,5-di trifluoromethyl phenylacetic acid ethyl ester, 2-3,5-bis trifluoromethyl phenyl acetic acid ethyl ester
42552891
ethyl 2-[3,5-bis(trifluoromethyl)phenyl]acetate
CCOC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
144632-97-3
C12H10F6O2
300.2
MFCD08689894
ICTVPOAERFKXSS-UHFFFAOYSA-N
ethyl 3,5-bis trifluoromethyl phenyl acetate, ethyl 3,5-bis trifluoromethyl phenylacetate, 3,5-bis trifluoromethyl phenylacetic acid ethyl ester, ethyl 2-3,5-bis trifluoromethyl phenyl acetate, ethyl 2-3,5-bis trifluoromethyl phenyl ethanoate, 3,5-di trifluoromethyl phenylacetic acid ethyl ester, 2-3,5-bis trifluoromethyl phenyl acetic acid ethyl ester
42552891
ethyl 2-[3,5-bis(trifluoromethyl)phenyl]acetate
CCOC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.417
Quantity 1 g
Formula Weight 300.2
Percent Purity 98%
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)phenylacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'