missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl 3-[[4-(Methylamino)-3-nitrobenzoyl]-(2-pyridyl)amino]propanoate, Mikromol™

Artikelnummer. 17972666
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17972666 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972666 Supplier Mikromol Supplier No. MM3522.100025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl 3-[[4-(Methylamino)-3-nitrobenzoyl]-(2-pyridyl)amino]propanoate
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Ethyl 3-[[4-(Methylamino)-3-nitrobenzoyl]-(2-pyridyl)amino]propanoate
CAS 429659-01-8
Analyt- eller komponentnavne Ethyl 3-[[4-(Methylamino)-3-nitrobenzoyl]-(2-pyridyl)amino]propanoate
Forsendelsestilstand Room Temperature
Molekylær formel C18 H20 N4 O5
Certificeringer/compliance ISO 9001
Synonym β-Alanine, N-[4-(methylamino)-3-nitrobenzoyl]-N-2-pyridinyl-, ethyl ester, Ethyl 3-[[4-(methylamino)-3-nitrobenzoyl](pyridin-2-yl)amino]propanoate
InChI formel InChI=1S/C18H20N4O5/c1-3-27-17(23)9-11-21(16-6-4-5-10-20-16)18(24)13-7-8-14(19-2)15(12-13)22(25)26/h4-8,10,12,19H,3,9,11H2,1-2H3
SMIL CCOC(=O)CCN(C(=O)c1ccc(NC)c(c1)[N+](=O)[O-])c2ccccn2
IUPAC navn ethyl 3-[[4-(methylamino)-3-nitrobenzoyl]-pyridin-2-ylamino]propanoate
Molekylvægt (g/mol) 372.38
Mængde 25 mg
Formel vægt 372.1434 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.