Få mere at vide
Ethyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Ethyl Ester), Mikromol™
Beskrivelse
Ethyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Ethyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Ethyl Ester) |
| CAS | 1458640-32-8 |
| Analyt- eller komponentnavne | Ethyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Ethyl Ester) |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C12 H18 N2 O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Carbidopa Imp. F (EP), (alphaS)-alpha-Hydrazinyl-3,4-dihydroxy-alpha-methyl-benzenepropanoic acid ethyl ester, Carbidopa Impurity F |
| InChI formel | InChI=1S/C12H18N2O4/c1-3-18-11(17)12(2,14-13)7-8-4-5-9(15)10(16)6-8/h4-6,14-16H,3,7,13H2,1-2H3/t12-/m0/s1 |
| SMIL | CCOC(=O)[C@](C)(Cc1ccc(O)c(O)c1)NN |
| IUPAC navn | ethyl (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoate |
| Vis mere |