Få mere at vide
Ethyl (2S)-2-Amino-4-phenylbutanoate Hydrochloride (L-Homophenylalanine Ethyl Ester Hydrochloride), Mikromol™
Beskrivelse
Ethyl (2S)-2-Amino-4-phenylbutanoate Hydrochloride (L-Homophenylalanine Ethyl Ester Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl (2S)-2-Amino-4-phenylbutanoate Hydrochloride |
| CAS | 90891-21-7 |
| Analyt- eller komponentnavne | Ethyl (2S)-2-Amino-4-phenylbutanoate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H17 N O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzenebutanoic acid, α-amino-, ethyl ester, hydrochloride (1:1), (αS)-, Benzenebutanoic acid, α-amino-, ethyl ester, hydrochloride, (S)-, Benzenebutanoic acid, α-amino-, ethyl ester, hydrochloride, (αS)- (9CI), Ethyl (S)-2-amino-4-phenylbutanoate hydrochloride, L-Homophenylalanine ethyl ester hydrochloride, Ethyl (2S)-2-Amino-4-phenylbutanoate Hydrochloride |
| InChI formel | InChI=1S/C12H17NO2.ClH/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10;/h3-7,11H,2,8-9,13H2,1H3;1H/t11-;/m0./s1 |
| SMIL | Cl.CCOC(=O)[C@@H](N)CCc1ccccc1 |
| IUPAC navn | ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride |
| Vis mere |