Få mere at vide
Ethyl (2S)-2-[(3S,5aS,9aS,10aS)-3-Methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl]pentanoate, Mikromol™
Artikelnummer. 17982816
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17982816 25 mg
1 options
Artikelnummer. 17982816 Leverandør Mikromol Leverandørnr. MM0926.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl (2S)-2-[(3S,5aS,9aS,10aS)-3-Methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl]pentanoate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-ethyl 2-{(3S,5aS,9aS,10aS)-3-methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl}pentanoate
CAS 129970-98-5
Analyt- eller komponentnavne (S)-ethyl 2-{(3S,5aS,9aS,10aS)-3-methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl}pentanoate
Forsendelsestilstand Room Temperature
Molekylær formel C19 H30 N2 O4
Certificeringer/compliance ISO 17025
Synonym (αS,3S,5aS,9aS,10aS)-decahydro-3-methyl-1,4-dioxo-α-propyl-Pyrazino[1,2-a]indole-2(1H)-acetic acid ethyl ester, Pyrazino[1,2-a]indole-2(1H)-acetic acid, decahydro-3-methyl-1,4-dioxo-α-propyl-, ethyl ester, [3S-[2(R*),3α,5aβ,9aβ,10aβ]]- (9CI), Perindopril Related Compound F (USP), Perindopril Related Compound F, Perindopril tert-Butylamine Impurity F, Perindopril Impurity F
InChI formel InChI=1S/C19H30N2O4/c1-4-8-15(19(24)25-5-2)20-12(3)17(22)21-14-10-7-6-9-13(14)11-16(21)18(20)23/h12-16H,4-11H2,1-3H3/t12-,13-,14-,15-,16-/m0/s1
SMIL CCC[C@H](N1[C@@H](C)C(=O)N2[C@H]3CCCC[C@H]3C[C@H]2C1=O)C(=O)OCC
IUPAC navn ethyl (2S)-2-[(3S,5aS,9aS,10aS)-3-methyl-1,4-dioxo-5a,6,7,8,9,9a,10,10a-octahydro-3H-pyrazino[1,2-a]indol-2-yl]pentanoate
Molekylvægt (g/mol) 350.45
Mængde 25 mg
Formel vægt 350.2206 g/mol