missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ethyl (2S)-2-[(3S,11aS)-3-Methyl-1,4-dioxo-1,3,4,6,11,11a-hexahydro-2H-pyrazino[1,2-b]isoquinolin-2-yl]-4-phenylbutanoat, Mikromol™
Description
Ethyl (2S)-2-[(3S,11aS)-3-Methyl-1,4-dioxo-1,3,4,6,11,11a-hexahydro-2H-pyrazino[1,2-b]isoquinolin-2-yl]-4-phenylbutanoat
Specifications
Specifications
| Kemisk navn eller materiale | Ethyl (2S)-2-[(3S,11aS)-3-Methyl-1,4-dioxo-1,3,4,6,11,11a-hexahydro-2H-pyrazino[1,2-b]isoquinolin-2-yl]-4-phenylbutanoate |
| CAS | 103733-49-9 |
| Analyt- eller komponentnavne | Ethyl (2S)-2-[(3S,11aS)-3-Methyl-1,4-dioxo-1,3,4,6,11,11a-hexahydro-2H-pyrazino[1,2-b]isoquinolin-2-yl]-4-phenylbutanoate |
| Forsendelsestilstand | Stuetemperatur |
| Molekylær formel | C25 H28 N2 O4 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Quinapril Hydrochloride Imp. D (EP), Quinapril Imp. D (EP), Quinapril USP Related Compound A, Quinapril USP RC A, Ethyl[3S-[2(R*),3a,11a beta]]-1,3,4,6,11,11a-hexahydro-3-methyl-1,4-dioxo-alpha-(2-phenylethyl)-2H-pyrazino[1,2-b]isoquinoline-2-acetate, Ethyl (2S)-2-[(3S,11aS)-3-Methyl-1,4-dioxo-1,3,4,6,11,11a-hexahydro-2H-pyrazino[1,2-b]isoquinolin-2-yl]-4-phenylbutanoate, Quinapril Related Compound A, Quinapril Hydrochloride Impurity D, Quinapril Impurity D |
| InChI formel | InChI=1S/C25H28N2O4/c1-3-31-25(30)21(14-13-18-9-5-4-6-10-18)27-17(2)23(28)26-16-20-12-8-7-11-19(20)15-22(26)24(27)29/h4-12,17,21-22H,3,13-16H2,1-2H3/t17-,21-,22-/m0/s1 |
| SMIL | CCOC(=O)[C@H](CCc1ccccc1)N2[C@@H](C)C(=O)N3Cc4ccccc4C[C@H]3C2=O |
| IUPAC navn | ethyl (2S)-2-[(3S,11aS)-3-methyl-1,4-dioxo-3,6,11,11a-tetrahydropyrazino[1,2-b]isoquinolin-2-yl]-4-phenylbutanoat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?