Få mere at vide
Ethyl (2RS)-2-(6-Chloro-9H-carbazol-2-yl)propanoate (Carprofen Ethyl Ester), Mikromol™
Beskrivelse
Ethyl (2RS)-2-(6-Chloro-9H-carbazol-2-yl)propanoate (Carprofen Ethyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Carprofen Ethyl Ester (Carprofen Impurity) |
| CAS | 52262-89-2 |
| Analyt- eller komponentnavne | Carprofen Ethyl Ester (Carprofen Impurity) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H16 Cl N O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Carprofen Imp. G (EP), Ethyl (2RS)-2-(6-Chloro-9H-carbazol-2-yl)propanoate (Carprofen Ethyl Ester), 6-Chloro-alpha-methyl-9H-carbazole-2-acetic acid ethyl ester, Carprofen Impurity G |
| InChI formel | InChI=1S/C17H16ClNO2/c1-3-21-17(20)10(2)11-4-6-13-14-9-12(18)5-7-15(14)19-16(13)8-11/h4-10,19H,3H2,1-2H3 |
| SMIL | CCOC(=O)C(C)c1ccc2c(c1)[nH]c3ccc(Cl)cc23 |
| IUPAC navn | ethyl 2-(6-chloro-9H-carbazol-2-yl)propanoate |
| Vis mere |