Learn More
Ethyl 2-(trifluoroacetyl)propionate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-(trifluoroacetyl)propionate |
| CAS | 344-00-3 |
| Molecular Formula | C7 H9 F3 O3 |
| Quantity | 100 mg |
| Synonym | Ethyl 2-methyl-4,4,4-trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate, Ethyl 4,4,4-trifluoro-2-methylacetoacetate, Ethyl (±)-2-methyl-4,4,4-trifluoroacetoacetate, Ethyl 2-methyl-4,4,4-trifluoroacetoacetate |
| InChI Formula | InChI=1S/C7H9F3O3/c1-3-13-6(12)4(2)5(11)7(8,9)10/h4H,3H2,1-2H3 |
| SMILES | CCOC(=O)C(C)C(=O)C(F)(F)F |
| IUPAC Name | ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate |
| Molecular Weight (g/mol) | 198.14 |
| Formula Weight | 198.05 |