missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate.
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
It is used as pharmaceutical intermediate.
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
Specifications
Melting Point | 145°C to 151°C |
Quantity | 250 mg |
Sensitivity | Moisture sensitive |
IUPAC Name | ethyl 2-methylpyridine-4-carboxylate hydrochloride |
Formula Weight | 201.65 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl 2-methylpyridine-4-carboxylate hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'