Learn More
Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
2037.00 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate.
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
It is used as pharmaceutical intermediate.
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
Specifications
Melting Point | 145°C to 151°C |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
IUPAC Name | ethyl 2-methylpyridine-4-carboxylate;hydrochloride |
Formula Weight | 201.65 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl 2-methylpyridine-4-carboxylate hydrochloride |