Learn More
Alfa Aesar™ Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate, tech. 90%
Brand:  Alfa Aesar™ H63971.MD
Additional Details : CAS Number : 2549-19-1 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15408616
  • 1138.66 DKK / 250mg
Estimated Shipment date
from Supplier 07-05-2018
Add to basket
Specifications
Chemical Identifiers
Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate
Technical Grade
MFCD00139509
2549-19-1
204.23
2-Methyl-imidazo[1,2-a]pyridine-3-carboxylic acid ethyl ester
Specifications
Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate
250mg
Technical Grade
90%
MFCD00139509
54° to 57°C
2549-19-1
Solid
204.23
2-Methyl-imidazo[1,2-a]pyridine-3-carboxylic acid ethyl ester