Få mere at vide
Ethyl 2-Methoxy-5-sulphamoylbenzoate, Mikromol™
Artikelnummer. 30052060
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052060 100 mg
30052262 25 mg
30462186 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30052060 Leverandør Mikromol Leverandørnr. MM0051.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 2-Methoxy-5-sulphamoylbenzoate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 2-Methoxy-5-sulphamoylbenzoate
CAS 33045-53-3
Analyt- eller komponentnavne Ethyl 2-Methoxy-5-sulphamoylbenzoate
Forsendelsestilstand Room Temperature
Molekylær formel C10 H13 N O5 S
Certificeringer/compliance ISO 17025
Synonym Ethyl 2-Methoxy-5-sulphamoylbenzoate, Sulpiride Imp. C (EP), Sulpiride Impurity C
InChI formel InChI=1S/C10H13NO5S/c1-3-16-10(12)8-6-7(17(11,13)14)4-5-9(8)15-2/h4-6H,3H2,1-2H3,(H2,11,13,14)
SMIL CCOC(=O)c1cc(ccc1OC)S(=O)(=O)N
IUPAC navn ethyl 2-methoxy-5-sulfamoylbenzoate
Molekylvægt (g/mol) 259.28
Mængde 100 mg
Formel vægt 259.0514 g/mol