missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl 2-Hydroxy-3-phenylpropanoate, TRC

Artikelnummer. 30379700
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379700 10 mg
30369141 100 mg
30371512 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30379700 Leverandør Toronto Research Chemicals Leverandørnr. TRCE76217310MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 15399-05-0
Molekylær formel C11H14O3
Molekylvægt (g/mol) 194.22
IUPAC navn ethyl 2-hydroxy-3-phenylpropanoate
SMIL CCOC(=O)C(O)Cc1ccccc1

Specifications

Kemisk navn eller materiale Ethyl 2-Hydroxy-3-phenylpropanoate
Mængde 10 mg
InChI formel InChI=1S/C11H14O3/c1-2-14-11(13)10(12)8-9-6-4-3-5-7-9/h3-7,10,12H,2,8H2,1H3
Formel vægt 194.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.