Få mere at vide
Ethyl 2-Hydroxy-3-phenylpropanoate, TRC
Artikelnummer. 30369141
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30369141 100 mg
30371512 50 mg
30379700 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30369141 Leverandør Toronto Research Chemicals Leverandørnr. TRCE762173100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15399-05-0
Molekylær formel C11H14O3
Molekylvægt (g/mol) 194.22
IUPAC navn ethyl 2-hydroxy-3-phenylpropanoate
SMIL CCOC(=O)C(O)Cc1ccccc1
Tekniske data
Kemisk navn eller materiale Ethyl 2-Hydroxy-3-phenylpropanoate
Mængde 100 mg
InChI formel InChI=1S/C11H14O3/c1-2-14-11(13)10(12)8-9-6-4-3-5-7-9/h3-7,10,12H,2,8H2,1H3
Formel vægt 194.0943