Få mere at vide
Ethyl 2-Hydrazinyl-2-oxoacetate, TRC
Artikelnummer. 30373360
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373360 100 mg
30370314 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30373360 Leverandør Toronto Research Chemicals Leverandørnr. TRCE918848100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 2-Hydrazinyl-2-oxoacetate
CAS 35196-48-6
Molekylær formel C4H8N2O3
Mængde 100 mg
InChI formel InChI=1S/C4H8N2O3/c1-2-9-4(8)3(7)6-5/h2,5H2,1H3,(H,6,7)
SMIL CCOC(=O)C(=O)NN
IUPAC navn ethyl 2-hydrazinyl-2-oxoacetate
Molekylvægt (g/mol) 132.11
Formel vægt 132.0535