Få mere at vide
Ethyl 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]amino]-2-oxoethyl]benzoate (Repaglinide Ethyl Ester), Mikromol™
Artikelnummer. 30051156
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051156 25 mg
30052139 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051156 Leverandør Mikromol Leverandørnr. MM0944.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]amino]-2-oxoethyl]benzoate (Repaglinide Ethyl Ester)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 2-Ethoxy-4-[2-(((1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]amino)-2-oxoethyl)-benzoate (Repaglinide Ethyl Ester)
CAS 147770-06-7
Analyt- eller komponentnavne Ethyl 2-Ethoxy-4-[2-(((1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]amino)-2-oxoethyl)-benzoate (Repaglinide Ethyl Ester)
Forsendelsestilstand Room Temperature
Molekylær formel C29 H40 N2 O4
Certificeringer/compliance ISO 9001
Synonym Ethyl 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]amino]-2-oxoethyl]-benzoate (Repaglinide Ethyl Ester), Repaglinide Imp. D (EP), 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]-benzoic acid ethyl ester, Benzoic acid, 2-ethoxy-4-[2-[[3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]-, ethyl ester, (S)-, (S)-2-Ethoxy-4-[[[N-[1-(2-piperidinophenyl)-3-methylbutyl]amino]carbonyl]methyl]benzoic acid ethyl ester, (S)-Ethyl 2-ethoxy-4-[[[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]amino]carbonyl]methyl]benzoate, Repaglinide Impurity D
InChI formel InChI=1S/C29H40N2O4/c1-5-34-27-19-22(14-15-24(27)29(33)35-6-2)20-28(32)30-25(18-21(3)4)23-12-8-9-13-26(23)31-16-10-7-11-17-31/h8-9,12-15,19,21,25H,5-7,10-11,16-18,20H2,1-4H3,(H,30,32)/t25-/m0/s1
SMIL CCOC(=O)c1ccc(CC(=O)N[C@@H](CC(C)C)c2ccccc2N3CCCCC3)cc1OCC
IUPAC navn ethyl 2-ethoxy-4-[2-[[(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]benzoate
Molekylvægt (g/mol) 480.64
Mængde 25 mg
Formel vægt 480.2988 g/mol