Få mere at vide
Ethyl 2-Ethoxy-1-((2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate, TRC
Artikelnummer. 30138755
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138755 500 mg
30137588 250 mg
30007597 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138755 Leverandør Toronto Research Chemicals Leverandørnr. TRCE678645500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1403474-70-3
Molekylær formel C27 H24 N4 O5
Molekylvægt (g/mol) 484.502
Synonym Ethyl 1-[[2'-(2,5-Dihydro-5-oxo-1,2,4-oxadiazol-3-yl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylate, Azilsartan Kamedoxomil Impurity, Azilsartan Impurity;
IUPAC navn ethyl 2-ethoxy-3-[[4-[2-(5-oxo-2H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMIL CCOC(=O)c1cccc2nc(OCC)n(Cc3ccc(cc3)c4ccccc4C5=NC(=O)ON5)c12
Tekniske data
Kemisk navn eller materiale Ethyl 2-Ethoxy-1-((2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate
Mængde 500 mg
InChI formel InChI=1S/C27H24N4O5/c1-3-34-25(32)21-10-7-11-22-23(21)31(26(28-22)35-4-2)16-17-12-14-18(15-13-17)19-8-5-6-9-20(19)24-29-27(33)36-30-24/h5-15H,3-4,16H2,1-2H3,(H,29,30,33)
Formel vægt 484.1747