missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloropyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number 15472329
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
89793-12-4
C7H7ClN2O2
186.595
MFCD09863164
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid ethyl ester, 2-chloro-5-ethoxycarbonyl pyrimidine, ethyl-2-chloropyrimidine-5-carboxylate, ethyl 2-chloro-pyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester, ethyl2-chloropyrimidine-5-carboxylate, acmc-209r2w, ksc663i8r, ethyl 2-chloranylpyrimidine-5-carboxylate, 2-chloro-5-pyrimidinecarboxylic acid ethyl ester
10487815
ethyl 2-chloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1)Cl
This item is not returnable. View return policy
89793-12-4
C7H7ClN2O2
186.595
MFCD09863164
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid ethyl ester, 2-chloro-5-ethoxycarbonyl pyrimidine, ethyl-2-chloropyrimidine-5-carboxylate, ethyl 2-chloro-pyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester, ethyl2-chloropyrimidine-5-carboxylate, acmc-209r2w, ksc663i8r, ethyl 2-chloranylpyrimidine-5-carboxylate, 2-chloro-5-pyrimidinecarboxylic acid ethyl ester
10487815
ethyl 2-chloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 60°C
Boiling Point 80°C (760 mmHg)
Quantity 250 mg
Beilstein 142651
Formula Weight 186.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 2-chloropyrimidine-5-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'