Learn More
Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate, 95%, Thermo Scientific Chemicals
1102.96 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
It is an active pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an active pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool dry ventilated place. Ensure proper ventilation.
It is an active pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool dry ventilated place. Ensure proper ventilation.
Specifications
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 200.62 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate |