Få mere at vide
Ethyl (2-Azidoethoxy-d4)acetoacetate, TRC
Artikelnummer. 30378353
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30378353 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378353 Leverandør Toronto Research Chemicals Leverandørnr. TRCE89955725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl (2-Azidoethoxy-d4)acetoacetate
CAS 1189691-91-5
Molekylær formel C8 2H4 H9 N3 O4
Mængde 25 mg
InChI formel InChI=1S/C8H13N3O4/c1-3-14-8(13)7(6(2)12)15-5-4-10-11-9/h7H,3-5H2,1-2H3/i4D2,5D2
SMIL [2H]C([2H])(OC(C(=O)C)C(=O)OCC)C([2H])([2H])N=[N+]=[N-]
IUPAC navn ethyl 2-(2-azido-1,1,2,2-tetradeuterioethoxy)-3-oxobutanoate
Molekylvægt (g/mol) 219.23
Formel vægt 219.1157