missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate, 97%, Thermo Scientific™

Catalog Number p-7032708
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
193537-14-3
C15H22N2O4S
326.411
MFCD05664039
XSXVOVXVHBSSSN-UHFFFAOYSA-N
ethyl 6-boc-2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3-carboxylate, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4,5-dihydrothieno 2,3-c pyridine-3,6 7h-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4h,5h,6h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4,7-dihydrothieno 2,3-c pyridine-3,6 5h-dicarboxylate, 2-amino-6-boc-4,5,6,7-tetrahydro-thieno 2,3-c pyridine-3-carboxylic acid ethyl ester, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert-butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4h,5h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, ethyl 2-amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylate, n-boc protected
2794740
6-O-tert-butyl 3-O-ethyl 2-amino-5,7-dihydro-4H-thieno[2,3-c]pyridine-3,6-dicarboxylate
CCOC(=O)C1=C(SC2=C1CCN(C2)C(=O)OC(C)(C)C)N
This item is not returnable. View return policy
193537-14-3
C15H22N2O4S
326.411
MFCD05664039
XSXVOVXVHBSSSN-UHFFFAOYSA-N
ethyl 6-boc-2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3-carboxylate, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4,5-dihydrothieno 2,3-c pyridine-3,6 7h-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4h,5h,6h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4,7-dihydrothieno 2,3-c pyridine-3,6 5h-dicarboxylate, 2-amino-6-boc-4,5,6,7-tetrahydro-thieno 2,3-c pyridine-3-carboxylic acid ethyl ester, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert-butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4h,5h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, ethyl 2-amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylate, n-boc protected
2794740
6-O-tert-butyl 3-O-ethyl 2-amino-5,7-dihydro-4H-thieno[2,3-c]pyridine-3,6-dicarboxylate
CCOC(=O)C1=C(SC2=C1CCN(C2)C(=O)OC(C)(C)C)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 151°C to 153°C
Quantity 250 mg
Formula Weight 326.42
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'