Learn More
Ethyl 2-amino-4-methylpyrimidine-5-carboxylate, 98%, BLD Pharm™
Product Code. 31449629
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31449629 10 g
31434864 5 g
31444723 100 g
31435985 25 g
31441969 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31449629 Supplier BLD Pharm Supplier No. BD12817210g
 Restricted Item
Forventet forsendelse: 25-11-2026
Specifications
CAS 81633-29-6
Molecular Formula C8H11N3O2
MDL Number MFCD00052622
Quantity 10 g
SMILES O=C(C1=CN=C(N)N=C1C)OCC
Molecular Weight (g/mol) 181.19
Percent Purity 98%
Chemical Name or Material Ethyl 2-amino-4-methylpyrimidine-5-carboxylate