missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-(5-methyl-2-furyl)thiophene-3-carboxylate, 96%, Thermo Scientific™

Catalog Number p-7032628
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
378196-87-3
C12H13NO3S
251.3
MFCD02654898
BGFHMNNBBDLFDU-UHFFFAOYSA-N
ethyl 2-amino-4-5-methylfuran-2-yl thiophene-3-carboxylate, 2-amino-4-5-methyl-furan-2-yl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4-5-methyl-2-furyl thiophene-3-carboxylate, ethyl 2-amino-4-5-methyl 2-furyl thiophene-3-carboxylate, ethyl 2-azanyl-4-5-methylfuran-2-yl thiophene-3-carboxylate, 2-amino-4-5-methyl-2-furanyl-3-thiophenecarboxylic acid ethyl ester, 2-amino-4-5-methyl-2-furyl thiophene-3-carboxylic acid ethyl ester
706051
ethyl 2-amino-4-(5-methylfuran-2-yl)thiophene-3-carboxylate
CCOC(=O)C1=C(SC=C1C2=CC=C(O2)C)N
This item is not returnable. View return policy
378196-87-3
C12H13NO3S
251.3
MFCD02654898
BGFHMNNBBDLFDU-UHFFFAOYSA-N
ethyl 2-amino-4-5-methylfuran-2-yl thiophene-3-carboxylate, 2-amino-4-5-methyl-furan-2-yl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4-5-methyl-2-furyl thiophene-3-carboxylate, ethyl 2-amino-4-5-methyl 2-furyl thiophene-3-carboxylate, ethyl 2-azanyl-4-5-methylfuran-2-yl thiophene-3-carboxylate, 2-amino-4-5-methyl-2-furanyl-3-thiophenecarboxylic acid ethyl ester, 2-amino-4-5-methyl-2-furyl thiophene-3-carboxylic acid ethyl ester
706051
ethyl 2-amino-4-(5-methylfuran-2-yl)thiophene-3-carboxylate
CCOC(=O)C1=C(SC=C1C2=CC=C(O2)C)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 91°C to 92°C
Quantity 250 mg
Formula Weight 251.31
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4-(5-methyl-2-furyl)thiophene-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'