Learn More
Alfa Aesar™ Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, 96%
Brand: Alfa Aesar™ H33319.MD
Additional Details : CAS Number : 193140-71-5 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
193140-71-5 | |
207.577 | |
MNFISCBCWBUTTQ-UHFFFAOYSA-N | |
45790935 | |
CCOC(=O)C(C(F)(F)F)N.Cl |
C5H9ClF3NO2 | |
MFCD14525479 | |
ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride, 2-amino-3,3,3-trifluoro-propionic acid ethyl ester hydrochloride, ethyl 3,3,3-trifluoroalaninate, ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, ethyl 3,3,3-trifluoroalaninate-hydrogen chloride 1/1 | |
ethyl 2-amino-3,3,3-trifluoropropanoate;hydrochloride |
Specifications
Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride | |
~149°C (decomposition) | |
193140-71-5 | |
MFCD14525479 | |
MNFISCBCWBUTTQ-UHFFFAOYSA-N | |
ethyl 2-amino-3,3,3-trifluoropropanoate;hydrochloride | |
45790935 | |
96% |
Hygroscopic | |
250mg | |
C5H9ClF3NO2 | |
ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride, 2-amino-3,3,3-trifluoro-propionic acid ethyl ester hydrochloride, ethyl 3,3,3-trifluoroalaninate, ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, ethyl 3,3,3-trifluoroalaninate-hydrogen chloride 1/1 | |
CCOC(=O)C(C(F)(F)F)N.Cl | |
207.577 | |
207.58 |