Få mere at vide
Ethyl 2-(6-Amino-2,3-dichlorobenzylamino)acetate Hydrochloride, Mikromol™
Beskrivelse
Ethyl 2-(6-Amino-2,3-dichlorobenzylamino)acetate Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 2-(6-Amino-2,3-dichlorobenzylamino)acetate Hydrochloride |
| CAS | 85325-12-8 |
| Analyt- eller komponentnavne | Ethyl 2-(6-Amino-2,3-dichlorobenzylamino)acetate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H14 Cl2 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Glycine, N-[(6-amino-2,3-dichlorophenyl)methyl]-, ethyl ester, hydrochloride (9CI), Anagrelide Related Compound A, Anagrelide Related Compound A as Hydrochloride |
| InChI formel | InChI=1S/C11H14Cl2N2O2.ClH/c1-2-17-10(16)6-15-5-7-9(14)4-3-8(12)11(7)13;/h3-4,15H,2,5-6,14H2,1H3;1H |
| SMIL | Cl.CCOC(=O)CNCc1c(N)ccc(Cl)c1Cl |
| IUPAC navn | ethyl 2-[(6-amino-2,3-dichlorophenyl)methylamino]acetate;hydrochloride |
| Vis mere |