Få mere at vide
Ethyl 2-(5,6-Dichloro-2-imino-1,2-dihydroquinazolin-3(4H)-yl)acetate Hydrobromide, Mikromol™
Artikelnummer. 30459879
Skift visning
Mængde:
25 mg
50 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459879 4 x 25 mg
17912836 50 mg
17953536 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459879 Leverandør Mikromol Leverandørnr. MM3250.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 2-(5,6-Dichloro-2-imino-1,2-dihydroquinazolin-3(4H)-yl)acetate Hydrobromide, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 2-(5,6-Dichloro-2-imino-1,2-dihydroquinazolin-3(4H)-yl)acetate Hydrobromide
CAS 70381-75-8
Analyt- eller komponentnavne Ethyl 2-(5,6-Dichloro-2-imino-1,2-dihydroquinazolin-3(4H)-yl)acetate Hydrobromide
Forsendelsestilstand Room Temperature
Molekylær formel C12 H13 Cl2 N3 O2 . Br H
Certificeringer/compliance ISO 17025
Synonym 3(4H)-Quinazolineacetic acid, 2-amino-5,6-dichloro-, ethyl ester, hydrobromide (1:1), 3(4H)-Quinazolineacetic acid, 2-amino-5,6-dichloro-, ethyl ester, monohydrobromide (9CI), Anagrelide Related Compound C (USP), Anagrelide Related Compound C as Hydrobromide
InChI formel InChI=1S/C12H13Cl2N3O2.BrH/c1-2-19-10(18)6-17-5-7-9(16-12(17)15)4-3-8(13)11(7)14;/h3-4H,2,5-6H2,1H3,(H2,15,16);1H
Løsningstype Neat
SMIL Br.CCOC(=O)CN1Cc2c(Cl)c(Cl)ccc2N=C1N
IUPAC navn ethyl 2-(2-amino-5,6-dichloro-4H-quinazolin-3-yl)acetate;hydrobromide
Molekylvægt (g/mol) 383.07
Mængde 4 x 25 mg
Formel vægt 380.9646 g/mol