Få mere at vide
Ethyl 2,4-Dichloro-5-sulphamoylbenzoate, Mikromol™
Artikelnummer. 30460479
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460479 4 x 25 mg
30051505 100 mg
30051882 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460479 Leverandør Mikromol Leverandørnr. MM0014.5000254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl 2,4-Dichloro-5-sulphamoylbenzoate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl 2,4-Dichloro-5-sulphamoylbenzoate
CAS 4793-25-3
Analyt- eller komponentnavne Ethyl 2,4-Dichloro-5-sulphamoylbenzoate
Forsendelsestilstand Room Temperature
Molekylær formel C9 H9 Cl2 N O4 S
Certificeringer/compliance ISO 17025
Synonym Ethyl 2,4-Dichloro-5-sulphamoylbenzoate
InChI formel InChI=1S/C9H9Cl2NO4S/c1-2-16-9(13)5-3-8(17(12,14)15)7(11)4-6(5)10/h3-4H,2H2,1H3,(H2,12,14,15)
Løsningstype Neat
SMIL CCOC(=O)c1cc(c(Cl)cc1Cl)S(=O)(=O)N
IUPAC navn ethyl 2,4-dichloro-5-sulfamoylbenzoate
Molekylvægt (g/mol) 298.14
Mængde 4 x 25 mg
Formel vægt 296.9629 g/mol