missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl 2-[3-Chloro-4-(3-pyrrolin-1-yl)phenyl]propionate, TRC

Artikelnummer. 30374780
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374780 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30374780 Leverandør Toronto Research Chemicals Leverandørnr. TRCE901995250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Ethyl 2-[3-Chloro-4-(3-pyrrolin-1-yl)phenyl]propionate
CAS 50537-11-6
Molekylær formel C15 H18 Cl N O2
Mængde 250 mg
Synonym 3-Chloro-4-(2,5-dihydro-1H-pyrrol-1-yl)-alpha-methyl-benzeneacetic Acid Ethyl Ester
InChI formel InChI=1S/C15H18ClNO2/c1-3-19-15(18)11(2)12-6-7-14(13(16)10-12)17-8-4-5-9-17/h4-7,10-11H,3,8-9H2,1-2H3
SMIL O=C(OCC)C(C1=CC=C(C(Cl)=C1)N2CC=CC2)C
IUPAC navn ethyl 2-[3-chloro-4-(2,5-dihydropyrrol-1-yl)phenyl]propanoate
Molekylvægt (g/mol) 279.76192
Formel vægt 279.1026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.