Learn More
Ethyl 2-(2-aminothiazol-4-yl)-2-oxoacetate, 97%, BLD Pharm™
Product Code. 31430137
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31430137 5 g
31424777 100 g
31417212 500 g
31420302 1 g
31420103 25 g
31423815 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31430137 Supplier BLD Pharm Supplier No. BD105665g
 Restricted Item
Forventet forsendelse: 25-11-2026
Specifications
CAS 64987-08-2
Molecular Formula C7H8N2O3S
MDL Number MFCD00010414
Quantity 5 g
SMILES O=C(OCC)C(C1=CSC(N)=N1)=O
Molecular Weight (g/mol) 200.22
Percent Purity 97%
Chemical Name or Material Ethyl 2-(2-aminothiazol-4-yl)-2-oxoacetate