Få mere at vide
Ethyl 2,2,2-Trifluoroethyl Carbonate, TRC
Artikelnummer. 30008388
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008388 1 g
30004066 250 mg
30007186 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008388 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9377331G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 156783-96-9
Molekylær formel C5 H7 F3 O3
Molekylvægt (g/mol) 172.102
Synonym 2,2,2-Trifluoroethyl Ethyl Carbonate;
IUPAC navn ethyl 2,2,2-trifluoroethyl carbonate
SMIL CCOC(=O)OCC(F)(F)F
Tekniske data
Kemisk navn eller materiale Ethyl 2,2,2-Trifluoroethyl Carbonate
Mængde 1 g
InChI formel InChI=1S/C5H7F3O3/c1-2-10-4(9)11-3-5(6,7)8/h2-3H2,1H3
Formel vægt 172.035