Få mere at vide
Ethyl-2,2,2-d3-malonic Acid, CDN
Artikelnummer. 30327269
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327269 0.25 g -
30316448 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327269 Leverandør CDN Leverandørnr. CDND51450.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl-2,2,2-d3-malonic acid
CAS 70907-93-6
Molekylær formel C5 D3 H5 O4
Mængde 0.25 g
Synonym Propanedioic acid, ethyl-2,2,2-d3- (9 CI), Ethyl-2,2,2-d3-malonic Acid, Ethyl-2,2,2-d3-propanedioic acid, 2-(Ethyl-2,2,2-d3)propanedioic acid, 1,1-Propanedicarboxylic-3,3,3-d3 acid, 2-(Ethyl-2,2,2-d3)malonic acid, Ethylmalonic Acid-D3 (ethyl-2,2,2-D3)
InChI formel InChI=1 S/C5H8O4/c1-2-3(4(6)7)5(8)9/h3H,2H2,1H3,(H,6,7)(H,8,9)/i1D3
SMIL [2 H]C([2 H])([2 H])CC(C(=O)O)C(=O)O
IUPAC navn 2-(2,2,2-trideuterioethyl)propanedioic acid
Molekylvægt (g/mol) 135.1331
Formel vægt 135.0611 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity