Få mere at vide
Ethyl-2,2,2-d3 Alcohol (95% w/w in H2O), CDN
Artikelnummer. 30312527
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30312527 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312527 Leverandør CDN Leverandørnr. CDND07521G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethyl-2,2,2-d3 Alcohol
CAS 1759-87-1
Opløsningsmiddel eller Matrix H2O
Molekylær formel C2 D3 H3 O
Mængde 1 g
Synonym Ethyl-2,2,2-d3 Alcohol, Ethan-2,2,2-d3-ol (9 CI), Ethyl-2,2,2-d3 alcohol (6 CI, 7 CI, 8 CI), 2,2,2-Trideuterioethanol, 2,2,2-Trideuteroethanol, Ethanol-2,2,2-d3, Ethanol-2,2,2-D3
InChI formel InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CO
IUPAC navn 2,2,2-trideuterioethanol
Molekylvægt (g/mol) 49.0869
Formel vægt 49.0607 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Koncentration eller sammensætning (efter analyt eller komponenter) 95 %