Få mere at vide
Ethyl (1RS,2SR)-2-(Methylamino)-1-phenylcyclohex-3-enecarboxylate Hydrochloride (Nortilidine Hydrochloride), Mikromol™
Beskrivelse
Ethyl (1RS,2SR)-2-(Methylamino)-1-phenylcyclohex-3-enecarboxylate Hydrochloride (Nortilidine Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Nortilidine HCl |
| CAS | 34596-11-7 |
| Analyt- eller komponentnavne | Nortilidine HCl |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H21 N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | trans-2-(Methylamino)-1-phenyl-3-cyclohexene-1-carboxylic acid ethyl ester hydrochloride, Ethyl (1RS,2SR)-2-(Methylamino)-1-phenylcyclohex-3-enecarboxylate Hydrochloride, Nortilidine Hydrochloride, Tilidine Hydrochloride Hemihydrate Impurity C as Hydrochloride, Tilidine Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C16H21NO2.ClH/c1-3-19-15(18)16(13-9-5-4-6-10-13)12-8-7-11-14(16)17-2;/h4-7,9-11,14,17H,3,8,12H2,1-2H3;1H/t14-,16+;/m0./s1 |
| SMIL | Cl.CCOC(=O)[C@]1(CCC=C[C@@H]1NC)c2ccccc2 |
| IUPAC navn | ethyl (1R,2S)-2-(methylamino)-1-phenylcyclohex-3-ene-1-carboxylate;hydrochloride |
| Vis mere |