Få mere at vide
Ethyl (1RS,2RS)-2-(Dimethylamino)-1-phenylcyclohex-3-enecarboxylate (cis-Tilidine), Mikromol
Beskrivelse
Ethyl (1RS,2RS)-2-(Dimethylamino)-1-phenylcyclohex-3-enecarboxylate (cis-Tilidine), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | cis-Tilidine (Ethyl (1RS,2RS)-2-(Dimethylamino)-1-phenylcyclohex-3-enecarboxylate) |
| CAS | 20380-56-7 |
| Analyt- eller komponentnavne | cis-Tilidine (Ethyl (1RS,2RS)-2-(Dimethylamino)-1-phenylcyclohex-3-enecarboxylate) |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C17 H23 N O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Tilidine Hydrochloride Hemihydrate Imp. A (EP), Tilidine Imp. A (EP), Ethyl (1RS,2RS)-2-(Dimethylamino)-1-phenylcyclohex-3-enecarboxylate, cis-Tilidine, 3-Cyclohexene-1-carboxylic acid, 2-(dimethylamino)-1-phenyl-, ethyl ester, (1R,2R)-rel-, 3-Cyclohexene-1-carboxylic acid, 2-(dimethylamino)-1-phenyl-, ethyl ester, cis-, 3-Cyclohexene-1-carboxylic acid, 2-(dimethylamino)-1-phenyl-, ethyl ester, stereoisomer (8CI), Ethyl cis-2-(dimethylamino)-1-phenyl-3-cyclohexene-1-carboxylate, Tilidine Hydrochloride Hemihydrate Impurity A, Tilidine Impurity A |
| InChI formel | InChI=1S/C17H23NO2/c1-4-20-16(19)17(14-10-6-5-7-11-14)13-9-8-12-15(17)18(2)3/h5-8,10-12,15H,4,9,13H2,1-3H3/t15-,17-/m1/s1 |
| Løsningstype | Neat |
| SMIL | CCOC(=O)[C@]1(CCC=C[C@H]1N(C)C)c2ccccc2 |
| Vis mere |