Få mere at vide
Ethyl 11,14-Diepoxyeicosanoate, TRC
Artikelnummer. 30133182
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133182 25 mg -
30132684 2.5 mg 2.5mg
17998554 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133182 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91600025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 355803-78-0
Molekylær formel C22 H40 O4
Molekylvægt (g/mol) 368.55
IUPAC navn ethyl 10-[3-[(3-pentyloxiran-2-yl)methyl]oxiran-2-yl]decanoate
SMIL CCCCCC1OC1CC2OC2CCCCCCCCCC(=O)OCC
Tekniske data
Kemisk navn eller materiale Ethyl 11,14-Diepoxyeicosanoate
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C22H40O4/c1-3-5-11-14-18-20(25-18)17-21-19(26-21)15-12-9-7-6-8-10-13-16-22(23)24-4-2/h18-21H,3-17H2,1-2H3
Formel vægt 368.2927
Procent renhed >95%