missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 1-benzylpiperidine-4-carboxylate, 97+%

Brand:  Alfa Aesar™ H52374.03

Additional Details : CAS Number : 24228-40-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439067

  • 202.00 DKK / 1g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

24228-40-8
247.338
ASQCOPJFYLJCGD-UHFFFAOYSA-N
90423
CCOC(=O)C1CCN(CC1)CC2=CC=CC=C2
C15H21NO2
MFCD00040748
ethyl n-benzylpiperidine-4-carboxylate, 1-benzylpiperidine-4-carboxylic acid ethyl ester, 1-benzyl-piperidine-4-carboxylic acid ethyl ester, ethyl 1-benzyl-4-piperidinecarboxylate, ethyl 1-benzylisonipecotate, 4-piperidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, 1-benzyl-4-piperidinecarboxylic acid ethyl ester, ethyl n-benzyl-isonipecotate, 1-benzyl-4-carboethoxypiperidine, 1-benzyl-piperidine-4-carboxylic acid-ethyl ester
ethyl 1-benzylpiperidine-4-carboxylate

Specifications

Ethyl 1-benzylpiperidine-4-carboxylate
24228-40-8
MFCD00040748
ASQCOPJFYLJCGD-UHFFFAOYSA-N
ethyl 1-benzylpiperidine-4-carboxylate
90423
≥97%
1g
C15H21NO2
ethyl n-benzylpiperidine-4-carboxylate, 1-benzylpiperidine-4-carboxylic acid ethyl ester, 1-benzyl-piperidine-4-carboxylic acid ethyl ester, ethyl 1-benzyl-4-piperidinecarboxylate, ethyl 1-benzylisonipecotate, 4-piperidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, 1-benzyl-4-piperidinecarboxylic acid ethyl ester, ethyl n-benzyl-isonipecotate, 1-benzyl-4-carboethoxypiperidine, 1-benzyl-piperidine-4-carboxylic acid-ethyl ester
CCOC(=O)C1CCN(CC1)CC2=CC=CC=C2
247.338
247.33
Videos
SDS Documents