Få mere at vide
Ethyl 1-Aminocyclopentanecarboxylate Hydrochloride, Mikromol™
Beskrivelse
Ethyl 1-Aminocyclopentanecarboxylate Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 1-Aminocyclopentanecarboxylate Hydrochloride |
| CAS | 22649-37-2 |
| Analyt- eller komponentnavne | Ethyl 1-Aminocyclopentanecarboxylate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H15 N O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Cyclopentanecarboxylic acid, 1-amino-, ethyl ester, hydrochloride (1:1), Cyclopentanecarboxylic acid, 1-amino-, ethyl ester, hydrochloride (6CI,8CI,9CI), Cycloleucine ethyl ester hydrochloride, Ethyl 1-Aminocyclopentanecarboxylate Hydrochloride |
| InChI formel | InChI=1S/C8H15NO2.ClH/c1-2-11-7(10)8(9)5-3-4-6-8;/h2-6,9H2,1H3;1H |
| SMIL | Cl.CCOC(=O)C1(N)CCCC1 |
| IUPAC navn | ethyl 1-aminocyclopentane-1-carboxylate;hydrochloride |
| Vis mere |