Få mere at vide
Ethyl-1,1-d2-amine HCl, CDN
Artikelnummer. 30327392
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327392 0.5 g 0.5g
30324698 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327392 Leverandør CDN Leverandørnr. CDND82890.5G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethylamine-1,1-D2 Hydrochloride
CAS 88877-94-5
Molekylær formel C2 D2 H5 N . Cl H
Mængde 0.5 g
Synonym Ethan-1,1-d2-amine, hydrochloride (9 CI), Ethan-1,1-d2-amine hydrochloride, Ethyl-1,1-d2-amine hydrochloride, 1,1-Dideuterioethylamine hydrochloride, Ethylamine-1,1-D2 hydrochloride
InChI formel InChI=1 S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1 H/i2D2;
SMIL Cl.[2 H]C([2 H])(C)N
IUPAC navn 1,1-dideuterioethanamine;hydrochloride
Molekylvægt (g/mol) 83.5569
Formel vægt 83.0471 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity