Få mere at vide
Ethoxzolamide, LoGiCal
Artikelnummer. 30462256
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30462256 5 mg
1 options
Artikelnummer. 30462256 Leverandør LoGiCal Leverandørnr. LGCFOR3082.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethoxzolamide
CAS 452-35-7
Analyt- eller komponentnavne Ethoxzolamide
Forsendelsestilstand Room Temperature
Molekylær formel C9 H10 N2 O3 S2
Certificeringer/compliance ISO 9001
Synonym 2-Benzothiazolesulfonamide, 6-ethoxy-, 6-Ethoxy-2-benzothiazolesulfonamide, 6-Ethoxyzolamide, Cardrase, Diuretic C, Ethamide, Ethoxyzolamide, Ethoxzolamide, Etoxzolamide, Glaucotensil, L 643786, NSC 10679, PNU 4191, Redupresin, U 4191
InChI formel InChI=1S/C9H10N2O3S2/c1-2-14-6-3-4-7-8(5-6)15-9(11-7)16(10,12)13/h3-5H,2H2,1H3,(H2,10,12,13)
Løsningstype Neat
SMIL CCOc1ccc2nc(sc2c1)S(=O)(=O)N
IUPAC navn 6-ethoxy-1,3-benzothiazole-2-sulfonamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 258.32
Mængde 5 mg
Formel vægt 258.0133 g/mol