Få mere at vide
Ethotoin, TRC
Artikelnummer. 30374561
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374561 500 mg
30378492 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30374561 Leverandør Toronto Research Chemicals Leverandørnr. TRCE890535500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 86-35-1
Molekylær formel C11 H12 N2 O2
Molekylvægt (g/mol) 204.23
Synonym 2,4-Imidazolidinedione, 3-ethyl-5-phenyl-, Hydantoin, 3-ethyl-5-phenyl- (6CI,7CI,8CI), (±)-Ethotoin, 1-Ethyl-2,5-dioxo-4-phenylimidazolidine, 3-Ethyl-5-phenyl-2,4-imidazolidinedione, 3-Ethyl-5-phenylhydantoin, 3-Ethyl-5-phenylimidazolidin-2,4-dione, AC-695, Accenon, Ethotoin, Peganone
IUPAC navn 3-ethyl-5-phenylimidazolidine-2,4-dione
SMIL CCN1C(=O)NC(C1=O)c2ccccc2
Tekniske data
Kemisk navn eller materiale Ethotoin
Mængde 500 mg
InChI formel InChI=1S/C11H12N2O2/c1-2-13-10(14)9(12-11(13)15)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,12,15)
Formel vægt 204.0899