Få mere at vide
Ethosuximide-d3, TRC
Artikelnummer. 30379285
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30379285 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379285 Leverandør Toronto Research Chemicals Leverandørnr. TRCE89044210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethosuximide-d3
CAS 1189703-33-0
Renhedsgrad noter GC
Molekylær formel C7H8D3NO2
Mængde 10 mg
Synonym Ethosuximide D3
InChI formel InChI=1S/C7H11NO2/c1-3-7(2)4-5(9)8-6(7)10/h3-4H2,1-2H3,(H,8,9,10)/i1D3
SMIL [2H]C([2H])([2H])CC1(C)CC(=O)NC1=O
Molekylvægt (g/mol) 144.19
Procent renhed >95